Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopinacolone, 97+%

Catalog No. AAA1065518
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 5469-26-1 | C6H11BrO | 179.057 g/mol

1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr

Specifications

Melting Point -10°C
Density 1.333
Boiling Point 189°C to 192°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.4655
Quantity 50 g
UN Number UN2810
Beilstein 506485
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 179.06
Percent Purity ≥97%
Chemical Name or Material 1-Bromopinacolone
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H318-H227-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 226-794-0
RTECSNumber EL7010000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.