Learn More
1-Bromopinacolone, 97+%, Thermo Scientific Chemicals
Description
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
Specifications
Melting Point | -10°C |
Density | 1.333 |
Boiling Point | 189°C to 192°C |
Flash Point | 75°C (167°F) |
Odor | Pungent |
Refractive Index | 1.4655 |
Quantity | 250 g |
UN Number | UN2810 |
Beilstein | 506485 |
Sensitivity | Light sensitive |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H318-H227-H315-H335 Toxic if inhaled. Causes serious eye damage. Combustible liquid. Causes skin irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S. |
EINECSNumber | 226-794-0 |
RTECSNumber | EL7010000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.