Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanesulfonic acid, sodium salt, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2386-54-1
C4H9NaO3S
160.17
MFCD00007540
XQCHMGAOAWZUPI-UHFFFAOYSA-M
sodium 1-butanesulfonate,sodium butane-1-sulfonate,1-butanesulfonic acid sodium salt,sodium butylsulfonate,1-butanesulfonic acid, sodium salt,sodium butane-1-sulphonate,sodium butanesulfonate,ipc-alks-4,1-butanesulfonic acid, sodium salt 1:1,acmc-1cdot
4096517
sodium;butane-1-sulfonate
CCCCS(=O)(=O)[O-].[Na+]
2386-54-1
C4H9NaO3S
160.17
MFCD00007540
XQCHMGAOAWZUPI-UHFFFAOYSA-M
sodium 1-butanesulfonate,sodium butane-1-sulfonate,1-butanesulfonic acid sodium salt,sodium butylsulfonate,1-butanesulfonic acid, sodium salt,sodium butane-1-sulphonate,sodium butanesulfonate,ipc-alks-4,1-butanesulfonic acid, sodium salt 1:1,acmc-1cdot
4096517
sodium;butane-1-sulfonate
CCCCS(=O)(=O)[O-].[Na+]
Packaging Glass bottle
Melting Point >300.0°C
Color White
Appearance (10% aq. soln.) clear to slightly hazy, colorless
Quantity 25g
Infrared Spectrum Authentic
Loss on Drying 2% max. (1 g, 105°C, 2 hrs)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)3SO3Na
Beilstein 04, 8
Solubility Information (10% in water) Clear to slightly hazy colorless
Formula Weight 160.17
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-Butanesulfonic acid, sodium salt
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 219-201-1