Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol,butanol,n-butanol,butyl alcohol,n-butyl alcohol,1-hydroxybutane,propylcarbinol,butyl hydroxide,propylmethanol,methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol,butanol,n-butanol,butyl alcohol,n-butyl alcohol,1-hydroxybutane,propylcarbinol,butyl hydroxide,propylmethanol,methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

High quality chemicals for laboratory and specialized industrial use

Formula Weight 74.12
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.81kg
Flash Point 37.00°C
Quantity 4L
Chemical Name or Material 1-Butanol