Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Steel Pail
Quantity 20L

 View more versions of this product

Catalog No. A399-20

  Check Availability
Add to cart

Custom packing/Bulk ordering

You can now order this item in custom quantities. Learn More

Specifications Description & Specifications

Chemical Identifiers

71-36-3
≥99.4 %
CH3CH2CH2CH2OH
MFCD00002964
1-Butanol
Certified ACS Reagent Grade, Suitable for fluorometric analysis
Liquid
C4H10O
74.12
Butyl alcohol
≥99.4%

Specifications

71-36-3
Liquid
C4H10O
74.12
1120; Colorless
Butyl alcohol
0.01% max.
Pass Test
Steel Pail
0.806 to 0.810 (at 25°C)
6.7mbar at 20°C
2.95 mPa/s at 20°C
117.6°C
−89°C
1-Butanol
Certified ACS Reagent Grade, Suitable for fluorometric analysis
Vapor Pressure: 6.7mbar at 20°C
≥99.4 %
CH3CH2CH2CH2OH
MFCD00002964
1.5°C max., 116.2° to 119.2°C
0.2% max.
2.6 (Air = 1.0)
Pass Test
0.005% max.
0.0008mEq/g max.
0.1% max.
Butanol
Colorless
20L
≥99.4%

Safety and Handling

SDS & Safety Documents

WARNING!

Emergency Overview
Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Alternative Products
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit