1-Butanol (Certified ACS), Fisher Chemical

Packaging Aluminum Can
Quantity 4L

 View more versions of this product

Catalog No. A399S-4

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 71-36-3
    Physical Form Liquid
    Assay Percent Range ≥99.4 %
    Molecular Formula C4H10O
    Linear Formula CH3CH2CH2CH2OH
    Formula Weight 74.12
    MDL Number MFCD00002964
    Synonym Butyl alcohol
    Chemical Name or Material 1-Butanol
    Assay ≥99.4%
    Grade Certified ACS Reagent Grade, Suitable for fluorometric analysis

    Specifications

    Boiling Point 117.6°C
    Melting Point −89°C
    Formula Weight 74.12
    Quantity 4L
    DOT Information DOT Class 3, : Flammable Liquid
    Flash Point 35°C (95°F)
    UN Number 1120; Colorless
    Physical Form Liquid
    Color Colorless
    Viscosity 2.95 mPa/s at 20°C
    Alpha Vector Butanol
    Density 2.6 (Air = 1.0)
    Vapor Pressure 6.7mbar at 20°C
    Molecular Formula C4H10O
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:2
    Flammability:3
    Instability:0
    Additional Information Vapor Pressure: 6.7mbar at 20°C
    Residue after Evaporation 0.005% max.
    Titratable Acid 0.0008mEq/g max.
    Cleanup Symbol Solvent
    Water 0.1% max.
    Assay Percent Range ≥99.4 %
    MDL Number MFCD00002964
    Boiling Range 1.5°C max., 116.2° to 119.2°C
    Synonym Butyl alcohol
    Health Hazard 2 WARNING!
    Identification Pass Test
    Packaging Aluminum Can
    Specific Gravity 0.806 to 0.810 (at 25°C)
    ChemAlert Storage Symbol Red
    Butyl Ether 0.2% max.
    Butyraldehyde 0.01% max.
    Fluorescence Background Pass Test
    Linear Formula CH3CH2CH2CH2OH
    CAS 71-36-3
    Chemical Name or Material 1-Butanol
    Assay ≥99.4%
    Grade Certified ACS Reagent Grade, Suitable for fluorometric analysis
       

    WARNING!

    Emergency Overview
    Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:2
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit