Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, HPLC Grade 99.4%, Thermo Scientific Chemicals

Catalog Number ac610020040
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for HPLC
TRUSTED_SUSTAINABILITY
Absorbance (1 cm cell vs water), 0.025 max. at 254nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 215nm
Assay Percent Range 99.50%
Linear Formula CH3(CH2)3OH
Beilstein 01, 367
Merck Index 15, 1542
Formula Weight 74.12
Percent Purity ≥99.4%
Grade HPLC
Density 2.6 (Air = 1.0)
Quantity 4 L
Identification Pass Test
Packaging EcoSafPak™
Refractive Index 1.3952 - 1.3992
Residue after Evaporation 0.005% max.
Titratable Acid 0.0008 meq/g max.
Vapor Pressure Vapor Pressure: 6.7mbar at 20°C
Water 0.1% max.
Chemical Name or Material 1-Butanol, 99.4%
Show More Show Less

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit