1-Butanol, HPLC Grade, 99%, Alfa Aesar™

Filtered through 0.2µm filters

Manufacturer: Alfa Aesar 22925M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA22925M1

 N/A
    N/A

This unit of measure is invalid. 


    Chemical Identifiers

    CAS 71-36-3
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula CH3(CH2)3OH
    Formula Weight 74.12
    MDL Number MFCD00002964
    Chemical Name or Material 1-Butanol
    Grade HPLC Grade

    Specifications

    Formula Weight 74.12
    Quantity 4 x 1L
    EINECS Number 200-751-6
    Physical Form Liquid
    Density 0.810
    Assay Percent Range 99%
    Molecular Formula CH3(CH2)3OH
    MDL Number MFCD00002964
    CAS 71-36-3
    Chemical Name or Material 1-Butanol
    Grade HPLC Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit