Alfa Aesar™ 1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%

Manufacturer: Alfa Aesar™ 32443K7

Quantity 4L

 View more versions of this product

Catalog No. AA32443K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71-36-3
    Linear Formula CH3(CH2)3OH
    Formula Weight 74.12
    MDL Number MFCD00002964
    UN Number UN1120
    Beilstein 969148
    Merck Index 14,1540
    Chemical Name or Material 1-Butanol
    Grade Spectrophotometric grade, Ultrapure

    Specifications

    Formula Weight 74.12
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number EO1400000
    Refractive Index 1.3990
    Boiling Point 117° to 118°C
    Color Colorless
    pH 7
    CAS 71-36-3
    UN Number UN1120
    Beilstein 969148
    Physical Form Liquid
    Merck Index 14,1540
    Health Hazard 1 Danger
    Flash Point 35° (95°F)
    Melting Point −90°C
    Quantity 4L
    MDL Number MFCD00002964
    Density 0.810g/mL
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
    TSCA TSCA
    Odor Alcohol-like
    Linear Formula CH3(CH2)3OH
    Health Hazard 2 GHS H Statement
    H318-H226-H302-H335-H336-H315
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    May cause respiratory irritation.
    May cause drowsiness or dizziness.
    Causes skin irritation.
    Chemical Name or Material 1-Butanol
    Assay ≥99.0%
    Grade Spectrophotometric grade, Ultrapure

    GHS H Statement
    H318-H226-H302-H335-H336-H315
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    May cause respiratory irritation.
    May cause drowsiness or dizziness.
    Causes skin irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit