Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%, Thermo Scientific Chemicals

Catalog Number AA32443M1
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

1-Butanol is primarily used as a solvent, as an intermediate in chemical synthesis, and as a fuel. It is also used as a paint thinner and a solvent in other coating applications where it is used as a relatively slow evaporating latent solvent in lacquers and ambient-cured enamels. It finds other uses such as a component of hydraulic and brake fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 7
Melting Point -90°C
Color Colorless
Density 0.810 g/mL
Boiling Point 117°C to 118°C
Quantity 4 x 1 L
Flash Point 35°C (95°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
Solubility Information Soluble in water.
Formula Weight 74.12
Percent Purity ≥99.0%
Grade Ultrapure
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Category H226-H302-H315-H318-H335-H336
Hazard Statement GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 200-751-6
RTECSNumber EO1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit