Alfa Aesar™ 1-Butenyl ethyl ether, cis + trans, 98%

Manufacturer: Alfa Aesar™ H2672022

Quantity 100g

 View more versions of this product

Catalog No. AAH2672022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 929-05-5
    Molecular Formula C6H12O
    Linear Formula CH3CH2CH=CHOCH2CH3
    Formula Weight 100.16
    MDL Number MFCD03427069
    UN Number UN3271
    Synonym 1-Ethoxy-1-butene
    Chemical Name or Material 1-Butenyl ethyl ether, Cis + Trans

    Specifications

    Formula Weight 100.16
    Molecular Formula C6H12O
    Linear Formula CH3CH2CH=CHOCH2CH3
    MDL Number MFCD03427069
    UN Number UN3271
    Synonym 1-Ethoxy-1-butene
    Health Hazard 1 Danger
    Boiling Point 94° to 95°C
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 100g
    Flash Point 6°C (43°F)
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    CAS 929-05-5
    Chemical Name or Material 1-Butenyl ethyl ether, Cis + Trans
    Assay 98%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit