Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2,3-dimethylimidazolium chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
98892-75-2
C9H17ClN2
188.699
MFCD03093291
HHHYPTORQNESCU-UHFFFAOYSA-M
1-butyl-2,3-dimethylimidazolium chloride,1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride,1-butyl-2,3-dimethylimidazol-1-ium chloride,3-butyl-1,2-dimethylimidazol-1-ium chloride,acmc-209sba,dsstox_cid_27882,dsstox_rid_82635,dsstox_gsid_47906,1-butyl-2,3-dimethylimidazoliumchloride,1-butyl-2,3-dime-thylimidazolium chloride
2734167
1-butyl-2,3-dimethylimidazol-3-ium;chloride
CCCCN1C=C[N+](=C1C)C.[Cl-]
98892-75-2
C9H17ClN2
188.699
MFCD03093291
HHHYPTORQNESCU-UHFFFAOYSA-M
1-butyl-2,3-dimethylimidazolium chloride,1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride,1-butyl-2,3-dimethylimidazol-1-ium chloride,3-butyl-1,2-dimethylimidazol-1-ium chloride,acmc-209sba,dsstox_cid_27882,dsstox_rid_82635,dsstox_gsid_47906,1-butyl-2,3-dimethylimidazoliumchloride,1-butyl-2,3-dime-thylimidazolium chloride
2734167
1-butyl-2,3-dimethylimidazol-3-ium;chloride
CCCCN1C=C[N+](=C1C)C.[Cl-]
Ionic liquid
Infrared Spectrum Authentic
Quantity 25g
Color White
Melting Point 87°C to 91°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 188.7
Physical Form Crystals
Percent Purity ≥98.0%
Chemical Name or Material 1-Butyl-2, 3-dimethylimidazolium chloride
Show More Show Less