Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 96%, Thermo Scientific™

Catalog Number AC356830100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
765910-73-4
C9H17N2·CF3O3S
302.32
MFCD03427624
KSOGGGZFEJTGPZ-UHFFFAOYSA-M
1-butyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-butyl-2,3-dimethylimidazolium triflate, dsstox_cid_29315, dsstox_rid_83431, dsstox_gsid_49356, ksc496s6h, 1-butyl-2,3-dimethylimidazol-1-ium triflate, 3-butyl-1,2-dimethylimidazol-1-ium triflate, 1-butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 1-butyl-2,3-dimethylimidazol-3-ium; trifluoromethanesulfonate
2734250
1-butyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]
765910-73-4
C9H17N2·CF3O3S
302.32
MFCD03427624
KSOGGGZFEJTGPZ-UHFFFAOYSA-M
1-butyl-2,3-dimethylimidazolium trifluoromethanesulfonate, 1-butyl-2,3-dimethylimidazolium triflate, dsstox_cid_29315, dsstox_rid_83431, dsstox_gsid_49356, ksc496s6h, 1-butyl-2,3-dimethylimidazol-1-ium triflate, 3-butyl-1,2-dimethylimidazol-1-ium triflate, 1-butyl-2,3-dimethylimidazolium trifluoromethansulfonate, 1-butyl-2,3-dimethylimidazol-3-ium; trifluoromethanesulfonate
2734250
1-butyl-2,3-dimethylimidazol-3-ium;trifluoromethanesulfonate
CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[O-]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Ionic liquid
TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.4480
Quantity 10 g
Formula Weight 302.32
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium trifluoromethansulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit