Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium (L)-lactate, 95+%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
878132-20-8
C11H20N2O3
228.292
MFCD03427627
CYFFKUDZLZREEX-UHFFFAOYSA-M
1-butyl-3-methyl-1h-imidazol-3-ium 2-hydroxypropanoate,1-butyl-3-methylimidazolium lactate
74941170
1-butyl-3-methylimidazol-3-ium;2-hydroxypropanoate
CCCCN1C=C[N+](=C1)C.CC(C(=O)[O-])O
878132-20-8
C11H20N2O3
228.292
MFCD03427627
CYFFKUDZLZREEX-UHFFFAOYSA-M
1-butyl-3-methyl-1h-imidazol-3-ium 2-hydroxypropanoate,1-butyl-3-methylimidazolium lactate
74941170
1-butyl-3-methylimidazol-3-ium;2-hydroxypropanoate
CCCCN1C=C[N+](=C1)C.CC(C(=O)[O-])O
Ionic liquid
Quantity 5g
Packaging Glass bottle
Color Yellow to Brown
Specific Rotation -4° to -6° (20°C, 589nm) (C=1, water)
Infrared Spectrum Authentic
Refractive Index 1.4910 to 1.4930 (20°C, 589nm)
Water 2% max. (K.F.)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95+%
Formula Weight 228.29
Physical Form Viscous Liquid
Percent Purity ≥95.0% (on dry substance)
Chemical Name or Material 1-Butyl-3-methylimidazolium (L)-lactate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC