Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-4-eth-1-ynylbenzene, 97%, Thermo Scientific™

Catalog Number BTB09904EA
Click to view available options
Quantity:
10 g
25 g
79887-09-5
C12H14
158.24
MFCD00173885
ZVWWYEHVIRMJIE-UHFFFAOYSA-N
4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene
2775132
1-butyl-4-ethynylbenzene
CCCCC1=CC=C(C=C1)C#C
79887-09-5
C12H14
158.24
MFCD00173885
ZVWWYEHVIRMJIE-UHFFFAOYSA-N
4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene
2775132
1-butyl-4-ethynylbenzene
CCCCC1=CC=C(C=C1)C#C
CAS Min % 96.5
CAS Max % 100.0
Density 0.906g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula HC2CC6H4(CH2)3CH3
Quantity 10 g
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 1-Butyl-4-eth-1-ynylbenzene
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit