Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Butyl isothiocyanate, 98+%

Manufacturer:  Alfa Aesar™ A1093506

Quantity 5g

 View more versions of this product

Catalog No. AAA1093506


  Check Availability
Add to cart

Specifications

Description

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
Description & Specifications

Chemical Identifiers

592-82-5
CH3(CH2)3NCS
MFCD00004824
906839
C5H9NS
115.2
UN2922
1-Butyl isothiocyanate

Specifications

592-82-5
5g
Pungent
CH3(CH2)3NCS
MFCD00004824
906839
≥98%
63° to 65°C (11mmHg)
66°C (150°F)
C5H9NS
115.2
UN2922
1-Butyl isothiocyanate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : NX8420500

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit