Alfa Aesar™ (1-Butyl)triphenylphosphonium bromide, 99%

Manufacturer: Alfa Aesar™ A1050422

Quantity 100g

 View more versions of this product

Catalog No. AAA1050422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1779-51-7
    Molecular Formula CH3(CH2)3P(C6H5)3Br
    Formula Weight 399.32
    MDL Number MFCD00011855
    UN Number UN3464
    Beilstein 3580224
    Chemical Name or Material (1-Butyl)triphenylphosphonium bromide

    Specifications

    Formula Weight 399.32
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011855
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
    TSCA TSCA
    UN Number UN3464
    Beilstein 3580224
    Health Hazard 1 Danger
    Melting Point 240° to 243°C
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number TA1855200
    Molecular Formula CH3(CH2)3P(C6H5)3Br
    CAS 1779-51-7
    Chemical Name or Material (1-Butyl)triphenylphosphonium bromide
    Assay 99%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit