Alfa Aesar™ 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%

Manufacturer: Alfa Aesar™ H2766103

Quantity 1g

 View more versions of this product

Catalog No. AAH2766103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85167-10-8
    Molecular Formula C14H25BO2
    Formula Weight 236.17
    MDL Number MFCD09037499
    Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol butylboronate
    Chemical Name or Material 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester

    Specifications

    Formula Weight 236.17
    Molecular Formula C14H25BO2
    Optical Rotation +25° (c=1 in Chloroform)
    MDL Number MFCD09037499
    CAS 85167-10-8
    Synonym (1S,2S,3R,5S)-(+)-2,3-Pinanediol butylboronate
    Quantity 1g
    Chemical Name or Material 1-Butylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit