Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylpyridinium chloride, ≥98%, Acros Organics

Click to view available options
Quantity:
25g
1124-64-7
C9H14ClN
171.668
POKOASTYJWUQJG-UHFFFAOYSA-M
1-butylpyridinium chloride,1-butylpyridin-1-ium chloride,n-butylpyridinium chloride,butylpyridinium chloride,dsstox_cid_11462,dsstox_rid_78879,dsstox_gsid_31462,unii-f8m63j351a,butylpyridiniumchloride,butylpyridine, chloride
2734171
1-butylpyridin-1-ium;chloride
CCCC[N+]1=CC=CC=C1.[Cl-]
1124-64-7
C9H14ClN
171.668
POKOASTYJWUQJG-UHFFFAOYSA-M
1-butylpyridinium chloride,1-butylpyridin-1-ium chloride,n-butylpyridinium chloride,butylpyridinium chloride,dsstox_cid_11462,dsstox_rid_78879,dsstox_gsid_31462,unii-f8m63j351a,butylpyridiniumchloride,butylpyridine, chloride
2734171
1-butylpyridin-1-ium;chloride
CCCC[N+]1=CC=CC=C1.[Cl-]
Ionic liquid
Quantity 25g
Color White
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 171.67
Physical Form Crystals
Percent Purity ≥98.0%
Chemical Name or Material 1-Butylpyridinium chloride