Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Carbamoylmethyl)pyridinium Chloride 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
41220-29-5
C7H9ClN2O
172.612
MFCD00032002
IMJBHWDMQIYCEI-UHFFFAOYSA-N
2756440
2-pyridin-1-ium-1-ylacetamide;chloride
C1=CC=[N+](C=C1)CC(=O)N.[Cl-]
41220-29-5
C7H9ClN2O
172.612
MFCD00032002
IMJBHWDMQIYCEI-UHFFFAOYSA-N
2756440
2-pyridin-1-ium-1-ylacetamide;chloride
C1=CC=[N+](C=C1)CC(=O)N.[Cl-]
Color Undesignated
Melting Point 213°C
Quantity 25g
Formula Weight 172.61
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material 1-(Carbamoylmethyl)pyridinium Chloride