Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
135540-11-3
C9H16ClN2·F6P
332.66
NHEGCUSBUWGOQM-UHFFFAOYSA-N
pyclu,1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate,chlorodipyrrolidinocarbenium hexafluorophosphate,1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v,1-chloro-1-pyrrolidinylmethylene pyrrolidinium,pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate,chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate,chloro-dipyrrolidinocarbenium hexafluorophosphate,pyclu chlorodipyrrolidinocarbenium hexafluorophosphate,1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
2736652
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)Cl.F[P-](F)(F)(F)(F)F
135540-11-3
C9H16ClN2·F6P
332.66
NHEGCUSBUWGOQM-UHFFFAOYSA-N
pyclu,1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate,chlorodipyrrolidinocarbenium hexafluorophosphate,1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v,1-chloro-1-pyrrolidinylmethylene pyrrolidinium,pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate,chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate,chloro-dipyrrolidinocarbenium hexafluorophosphate,pyclu chlorodipyrrolidinocarbenium hexafluorophosphate,1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
2736652
1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)Cl.F[P-](F)(F)(F)(F)F
Coupling reagent
Melting Point 152°C to 156°C
Infrared Spectrum Authentic
Packaging Glass bottle
Elemental Analysis 31.68 to 33.31% C, 8.21 to 8.63% N
Quantity 5g
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 332.66
Physical Form Powder or Solid
Percent Purity 98%
Chemical Name or Material 1-(chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning