Alfa Aesar™ 1-Chloro-2,4-bis(trifluoromethyl)benzene, 97%

Manufacturer: Alfa Aesar™ B2273406

Quantity 5g

 View more versions of this product

Catalog No. AAB2273406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 327-76-4
    Molecular Formula C8H3ClF6
    Formula Weight 248.56
    MDL Number MFCD01631638
    UN Number UN1993
    Beilstein 1978012
    Synonym 2,4-Bis(trifluoromethyl)chlorobenzene; 4-Chloro-1,3-bis(trifluoromethyl)benzene
    Chemical Name or Material 1-Chloro-2,4-bis(trifluoromethyl)benzene

    Specifications

    Formula Weight 248.56
    Molecular Formula C8H3ClF6
    UN Number UN1993
    Melting Point −59°C
    MDL Number MFCD01631638
    CAS 327-76-4
    Beilstein 1978012
    Synonym 2,4-Bis(trifluoromethyl)chlorobenzene; 4-Chloro-1,3-bis(trifluoromethyl)benzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 59°C (138°F)
    Boiling Point 150° to 154°C
    Quantity 5g
    Chemical Name or Material 1-Chloro-2,4-bis(trifluoromethyl)benzene
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit