Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-2,4-bis(trifluoromethyl)benzene, 97%

Click to view available options
Quantity:
1g
5g
327-76-4
C8H3ClF6
248.552
MFCD01631638
XIVDTLKMHDHQCB-UHFFFAOYSA-N
1-chloro-2,4-bis trifluoromethyl benzene,2,4-bis trifluoromethyl chlorobenzene,2,4-bis-trifluoromethyl chlorobenzene,1,3-bis trifluoromethyl-4-chlorobenzene,1-chloro-2,4-bis-trifluoromethyl benzene,2,4-bis trifluoromethyl-1-chlorobenzene,pubchem8495,acmc-20a3u6,1,3-bis trifluoromethyl-4-chlorobenzen,1-chloro-2,4-bis-trifluoromethyl-benzene
2773209
1-chloro-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)Cl
327-76-4
C8H3ClF6
248.552
MFCD01631638
XIVDTLKMHDHQCB-UHFFFAOYSA-N
1-chloro-2,4-bis trifluoromethyl benzene,2,4-bis trifluoromethyl chlorobenzene,2,4-bis-trifluoromethyl chlorobenzene,1,3-bis trifluoromethyl-4-chlorobenzene,1-chloro-2,4-bis-trifluoromethyl benzene,2,4-bis trifluoromethyl-1-chlorobenzene,pubchem8495,acmc-20a3u6,1,3-bis trifluoromethyl-4-chlorobenzen,1-chloro-2,4-bis-trifluoromethyl-benzene
2773209
1-chloro-2,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)Cl
Refractive Index 1.414
Density 1.52
Melting Point -59°C
Flash Point 59°C (138°F)
Quantity 1g
Boiling Point 150°C to 154°C
UN Number UN1993
Beilstein 1978012
Formula Weight 248.56
Percent Purity 97%
Chemical Name or Material 1-Chloro-2,4-bis(trifluoromethyl)benzene
Show More Show Less

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures