Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-2,5-bis(trifluoromethyl)benzene, 98%

Click to view available options
Quantity:
1g
5g
25g
328-91-6
C8H3ClF6
248.552
MFCD00632388
IDNVTCBYGYOPKJ-UHFFFAOYSA-N
2,5-bis trifluoromethyl chlorobenzene,2-chloro-1,4-bis trifluoromethyl benzene,1-chloro-2,5-bis trifluoromethyl benzene,1,4-bis trifluoromethyl-2-chlorobenzene,1-chloro-2,5-bis trifluoromethyl benzene,2,5-ditrifluoromethyl-chlorobenzene,2-chlor-1,4-bis-trifluormethyl-benzol,2-chloro-1,4-bis trifluoromethyl benzene #,benzene, 1-chloro-2,5-bis trifluoromethyl,benzene, 2-chloro-1,4-bis trifluoromethyl
619189
2-chloro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F
328-91-6
C8H3ClF6
248.552
MFCD00632388
IDNVTCBYGYOPKJ-UHFFFAOYSA-N
2,5-bis trifluoromethyl chlorobenzene,2-chloro-1,4-bis trifluoromethyl benzene,1-chloro-2,5-bis trifluoromethyl benzene,1,4-bis trifluoromethyl-2-chlorobenzene,1-chloro-2,5-bis trifluoromethyl benzene,2,5-ditrifluoromethyl-chlorobenzene,2-chlor-1,4-bis-trifluormethyl-benzol,2-chloro-1,4-bis trifluoromethyl benzene #,benzene, 1-chloro-2,5-bis trifluoromethyl,benzene, 2-chloro-1,4-bis trifluoromethyl
619189
2-chloro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Cl)C(F)(F)F
Applications
It can be used as a synthetic building block.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Refractive Index 1.4135
Quantity 25g
Boiling Point 147°C to 149°C
Density 1.504g/mL
Beilstein 1978018
Formula Weight 248.55
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Chloro-2,5-bis(trifluoromethyl)benzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning