Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-2-cycloheptylethane, 96%

Click to view available options
Quantity:
5g
25g
84226-37-9
C9H17Cl
160.685
MFCD01632144
MPSIMZVBCYTZHR-UHFFFAOYSA-N
1-chloro-2-cycloheptylethane,2-chloroethyl cycloheptane,2-cycloheptylethyl chloride
20187818
2-chloroethylcycloheptane
C1CCCC(CC1)CCCl
84226-37-9
C9H17Cl
160.685
MFCD01632144
MPSIMZVBCYTZHR-UHFFFAOYSA-N
1-chloro-2-cycloheptylethane,2-chloroethyl cycloheptane,2-cycloheptylethyl chloride
20187818
2-chloroethylcycloheptane
C1CCCC(CC1)CCCl
Quantity 5g
Formula Weight 160.69
Percent Purity 96%
Chemical Name or Material 1-Chloro-2-cycloheptylethane

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning