Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-2-iodobenzene, 99%

Click to view available options
Quantity:
10g
25g
100g
615-41-8
C6H4ClI
238.452
MFCD00001033
MPEOPBCQHNWNFB-UHFFFAOYSA-N
2-chloroiodobenzene,o-chloroiodobenzene,o-iodochlorobenzene,2-iodochlorobenzene,benzene, 1-chloro-2-iodo,2-chlorophenyl iodide,2-chloro-1-iodobenzene,1-chloro-2-iodo-benzene,chlorjodbenzol,iodochlorobenzene
11993
1-chloro-2-iodobenzene
C1=CC=C(C(=C1)Cl)I
615-41-8
C6H4ClI
238.452
MFCD00001033
MPEOPBCQHNWNFB-UHFFFAOYSA-N
2-chloroiodobenzene,o-chloroiodobenzene,o-iodochlorobenzene,2-iodochlorobenzene,benzene, 1-chloro-2-iodo,2-chlorophenyl iodide,2-chloro-1-iodobenzene,1-chloro-2-iodo-benzene,chlorjodbenzol,iodochlorobenzene
11993
1-chloro-2-iodobenzene
C1=CC=C(C(=C1)Cl)I
Applications
1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2′-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place. Keep the container tightly closed.
Quantity 100g
Refractive Index 1.6345
Boiling Point 108°C to 110°C (15mmHg)
Density 1.954
Sensitivity Light sensitive
Flash Point 111°C (231°F)
Melting Point 1°C
Beilstein 1905116
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 238.46
Percent Purity 99%
Chemical Name or Material 1-Chloro-2-iodobenzene
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-423-4

TSCA : Yes

Recommended Storage : Ambient temperatures