Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3,4-dinitrobenzene, 90%, ACROS Organics™

Click to view available options
Quantity:
50g
100g
250g
Packaging:
Glass Bottle
610-40-2
C6H3ClN2O4
202.55
QVQSOXMXXFZAKU-UHFFFAOYSA-N
1-chloro-3,4-dinitrobenzene,3,4-dinitrochlorobenzene,benzene, 4-chloro-1,2-dinitro,unii-z29b1f274e,1,2-dinitro-4-chlorobenzene,1,2-dino2 4-cl benzene,4-chlordinitrobenzol,acmc-209mog,4-chloro-1,2-dinitrobenzen,ghl.pd_mitscher_leg0.928
33097
4-chloro-1,2-dinitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]
610-40-2
C6H3ClN2O4
202.55
QVQSOXMXXFZAKU-UHFFFAOYSA-N
1-chloro-3,4-dinitrobenzene,3,4-dinitrochlorobenzene,benzene, 4-chloro-1,2-dinitro,unii-z29b1f274e,1,2-dinitro-4-chlorobenzene,1,2-dino2 4-cl benzene,4-chlordinitrobenzol,acmc-209mog,4-chloro-1,2-dinitrobenzen,ghl.pd_mitscher_leg0.928
33097
4-chloro-1,2-dinitrobenzene
C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]
Packaging Glass Bottle
Infrared Spectrum Authentic
Refractive Index 1.5865 to 1.5885 (20°C, 589nm)
Specific Gravity 1.68
Flash Point 194°C
Color Brown-Yellow
Quantity 250g
Density 1.6800g/mL
Boiling Point 315.0°C
Melting Point 36.0°C
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula ClC6H3(NO2)2
Beilstein 05,262
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 202.55
Physical Form Oily Liquid After Melting
Percent Purity 90%
Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 210-223-7