Alfa Aesar™ 1-Chloro-3,4-dinitrobenzene, tech. 90%

Manufacturer: Alfa Aesar™ B2277318

Quantity 50g

 View more versions of this product

Catalog No. AAB2277318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 610-40-2
    Molecular Formula C6H3ClN2O4
    Formula Weight 202.55
    MDL Number MFCD00007212
    UN Number UN1577
    Beilstein 521564
    Synonym 3,4-Dinitrochlorobenzene
    Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
    Grade Technical Grade

    Specifications

    Formula Weight 202.55
    MDL Number MFCD00007212
    UN Number UN1577
    Flash Point >110°C (230°F)
    Quantity 50g
    TSCA TSCA
    Molecular Formula C6H3ClN2O4
    Beilstein 521564
    Synonym 3,4-Dinitrochlorobenzene
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, LIQUID.
    Boiling Point 14° to 16°C (4mmHg)
    Melting Point ≈20° to 25°C
    CAS 610-40-2
    Health Hazard 1 Danger
    Chemical Name or Material 1-Chloro-3,4-dinitrobenzene
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit