Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-3,5-dimethoxybenzene, 98%

Click to view available options
Quantity:
5g
25g
100g
7051-16-3
C8H9ClO2
172.608
MFCD00008382
WQHNWJBSROXROL-UHFFFAOYSA-N
5-chloro-1,3-dimethoxybenzene,3,5-dimethoxychlorobenzene,benzene, 1-chloro-3,5-dimethoxy,5-chlororesorcinol dimethyl ether,1,3-dimethoxy-5-chlorobenzene,pubchem3042,benzene,5-dimethoxy,acmc-209oev,wln: gr co1 eo1,3,5-dimethoxyphenyl chloride
81502
1-chloro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Cl)OC
7051-16-3
C8H9ClO2
172.608
MFCD00008382
WQHNWJBSROXROL-UHFFFAOYSA-N
5-chloro-1,3-dimethoxybenzene,3,5-dimethoxychlorobenzene,benzene, 1-chloro-3,5-dimethoxy,5-chlororesorcinol dimethyl ether,1,3-dimethoxy-5-chlorobenzene,pubchem3042,benzene,5-dimethoxy,acmc-209oev,wln: gr co1 eo1,3,5-dimethoxyphenyl chloride
81502
1-chloro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Cl)OC
Melting Point 32°C to 36°C
Boiling Point 234°C
Quantity 100g
Flash Point 112°C (233°F)
Beilstein 2327304
Formula Weight 172.61
Percent Purity 98%
Chemical Name or Material 1-Chloro-3,5-dimethoxybenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CZ0320000