Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-3-iodobenzene, 98%

Click to view available options
Quantity:
5g
25g
100g
625-99-0
C6H4ClI
238.452
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene,m-chloroiodobenzene,benzene, 1-chloro-3-iodo,3-iodochlorobenzene,3-chloro-1-iodobenzene,1-chloro-3-iodo-benzene,m-chlorophenyl iodide,m-iodochlorobenzene,metachloroiodobenzene,3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
C1=CC(=CC(=C1)I)Cl
625-99-0
C6H4ClI
238.452
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene,m-chloroiodobenzene,benzene, 1-chloro-3-iodo,3-iodochlorobenzene,3-chloro-1-iodobenzene,1-chloro-3-iodo-benzene,m-chlorophenyl iodide,m-iodochlorobenzene,metachloroiodobenzene,3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
C1=CC(=CC(=C1)I)Cl
Applications
1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 67.2 mg/L.

Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Density 1.927
Sensitivity Light sensitive
Refractive Index 1.631
Quantity 25g
Boiling Point 228°C
Flash Point 102°C (216°F)
Beilstein 1904539
Solubility Information Soluble in water 67.2mg/L.
Formula Weight 238.46
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodobenzene
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-920-6

TSCA : Yes

Recommended Storage : Ambient temperatures