Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-iodobenzene (stabilized with Copper chip) 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
625-99-0
C6H4ClI
238.45
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
ClC1=CC=CC(I)=C1
625-99-0
C6H4ClI
238.45
MFCD00001046
JMLWXCJXOYDXRN-UHFFFAOYSA-N
3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
12269
1-chloro-3-iodobenzene
ClC1=CC=CC(I)=C1
Quantity 25g
Color Undesignated
Boiling Point 228°C
Formula Weight 238.45
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1-Chloro-3-iodobenzene (stabilized with Copper chip)