Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-3-iodopropane, 98%, stabilized, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6940-76-7
C3H6ClI
204.44
MFCD00001096
SFOYQZYQTQDRIY-UHFFFAOYSA-N
propane, 1-chloro-3-iodo,3-chloropropyl iodide,trimethylene chloroiodide,1-chloro-3-iodo-propane,3-chloroiodopropane,3-chloro-iodopropane,1-chloro-3iodopropane,1-choro-3-iodopropane,3-chloro-l-iodopropane,1-iodo-3-chloropropane
81363
1-chloro-3-iodopropane
C(CCl)CI
6940-76-7
C3H6ClI
204.44
MFCD00001096
SFOYQZYQTQDRIY-UHFFFAOYSA-N
propane, 1-chloro-3-iodo,3-chloropropyl iodide,trimethylene chloroiodide,1-chloro-3-iodo-propane,3-chloroiodopropane,3-chloro-iodopropane,1-chloro-3iodopropane,1-choro-3-iodopropane,3-chloro-l-iodopropane,1-iodo-3-chloropropane
81363
1-chloro-3-iodopropane
C(CCl)CI
Specific Gravity 1.9
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 170.0°C to 172.0°C
Density 1.9000g/mL
Flash Point >110°C
Stabilizer copper
Refractive Index 1.5463 to 1.5483
Quantity 100g
Color Orange to Pink or Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula Cl(CH2)3I
Beilstein 01, 114
Formula Weight 204.44
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodopropane, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-088-8

TSCA : TSCA