Alfa Aesar™ 1-Chloro-3-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ B234230B

Quantity 1000g

 View more versions of this product

Catalog No. AAB234230B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121-73-3
    Molecular Formula C6H4ClNO2
    Formula Weight 157.56
    MDL Number MFCD00007202
    UN Number UN1578
    Beilstein 1907947
    Merck Index 14,2151
    Synonym 3-Nitrochlorobenzene
    Chemical Name or Material 1-Chloro-3-nitrobenzene

    Specifications

    Formula Weight 157.56
    TSCA TSCA
    Melting Point 43° to 47°C
    Quantity 1000g
    Flash Point 124°C (255°F)
    Molecular Formula C6H4ClNO2
    MDL Number MFCD00007202
    Beilstein 1907947
    Merck Index 14,2151
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H311-H373-H302
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORONITROBENZENES, SOLID
    Boiling Point 235° to 236°C
    CAS 121-73-3
    UN Number UN1578
    Synonym 3-Nitrochlorobenzene
    Health Hazard 3 GHS P Statement
    P260-P280-P361-P301+P312-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Remove/Take off immediately all contaminated clothing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CZ0940000
    Chemical Name or Material 1-Chloro-3-nitrobenzene
    Assay 98%

    GHS H Statement
    H311-H373-H302
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.

    GHS P Statement
    P260-P280-P361-P301+P312-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Remove/Take off immediately all contaminated clothing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit