Alfa Aesar™ 1-Chloro-4-difluoromethyl-2-iodobenzene, 97%

Manufacturer: Alfa Aesar™ H3410803

Quantity 1g

 View more versions of this product

Catalog No. AAH3410803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1261737-24-9
    Molecular Formula C7H4ClF2I
    Formula Weight 288.46
    MDL Number MFCD18392845
    Chemical Name or Material 1-Chloro-4-difluoromethyl-2-iodobenzene

    Specifications

    Formula Weight 288.46
    Molecular Formula C7H4ClF2I
    MDL Number MFCD18392845
    CAS 1261737-24-9
    Quantity 1g
    Chemical Name or Material 1-Chloro-4-difluoromethyl-2-iodobenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit