Alfa Aesar™ 1-Chloro-4-fluoro-2-nitrobenzene, 98+%

Manufacturer: Alfa Aesar™ A1140014

Quantity 25g

 View more versions of this product

Catalog No. AAA1140014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 345-17-5
    Molecular Formula C6H3ClFNO2
    Formula Weight 175.55
    MDL Number MFCD00084853
    UN Number UN2811
    Beilstein 2094191
    Synonym 2-Chloro-5-fluoronitrobenzene
    Chemical Name or Material 1-Chloro-4-fluoro-2-nitrobenzene

    Specifications

    Formula Weight 175.55
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H3ClFNO2
    Health Hazard 2 GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 37° to 39°C
    Odor Characteristic
    CAS 345-17-5
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Flash Point >110°C (230°F)
    MDL Number MFCD00084853
    UN Number UN2811
    Beilstein 2094191
    Synonym 2-Chloro-5-fluoronitrobenzene
    Health Hazard 1 Danger
    Boiling Point 237° to 239°C
    Quantity 25g
    Chemical Name or Material 1-Chloro-4-fluoro-2-nitrobenzene
    Assay ≥98%

    GHS H Statement
    H301-H311-H373-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P301+P310-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit