Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloro-4-fluoro-2-nitrobenzene, 98+%

Manufacturer:  Alfa Aesar™ A1140003

Quantity 1g

 View more versions of this product

Catalog No. AAA1140003


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

345-17-5
175.55
UN2811
2-Chloro-5-fluoronitrobenzene
C6H3ClFNO2
MFCD00084853
2094191
1-Chloro-4-fluoro-2-nitrobenzene

Specifications

>110°C (230°F)
37° to 39°C
345-17-5
C6H3ClFNO2
MFCD00084853
2094191
1-Chloro-4-fluoro-2-nitrobenzene
237° to 239°C
1g
Characteristic
175.55
UN2811
2-Chloro-5-fluoronitrobenzene
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H373-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit