Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-4-iodobenzene 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
637-87-6
C6H4ClI
238.46
MFCD00001053
GWQSENYKCGJTRI-UHFFFAOYSA-N
4-chloroiodobenzene,p-chloroiodobenzene,4-iodochlorobenzene,p-iodochlorobenzene,benzene, 1-chloro-4-iodo,1-chloro-4-iodo-benzene,p-chlorjodbenzol,4-chloro iodobenzene,4-chloro-iodobenzene,4-iodo-chlorobenzene
12510
1-chloro-4-iodobenzene
C1=CC(=CC=C1Cl)I
637-87-6
C6H4ClI
238.46
MFCD00001053
GWQSENYKCGJTRI-UHFFFAOYSA-N
4-chloroiodobenzene,p-chloroiodobenzene,4-iodochlorobenzene,p-iodochlorobenzene,benzene, 1-chloro-4-iodo,1-chloro-4-iodo-benzene,p-chlorjodbenzol,4-chloro iodobenzene,4-chloro-iodobenzene,4-iodo-chlorobenzene
12510
1-chloro-4-iodobenzene
C1=CC(=CC=C1Cl)I
Quantity 100g
Flash Point 108°C
Color White to Beige
Boiling Point 226°C to 227°C
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 52°C to 55°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H4I
Beilstein 05, 221
Formula Weight 238.46
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98.5%
Chemical Name or Material 1-Chloro-4-iodobenzene, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-305-5

TSCA : TSCA