Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-4-iodobutane, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
10297-05-9
C4H8ClI
218.462
MFCD00039415
JXOSPTBRSOYXGC-UHFFFAOYSA-N
butane, 1-chloro-4-iodo,tetramethylene chloroiodide,4-chlorobutyl iodide,4-iodobutyl chloride,1-chloro4-iodobutane,4-chloro-1-iodobutane,4-iodo-1-chlorobutane,1-iodo-4-chlorobutane,acmc-1buxg,1-chloro-4-iodo-butane
82527
1-chloro-4-iodobutane
C(CCI)CCl
10297-05-9
C4H8ClI
218.462
MFCD00039415
JXOSPTBRSOYXGC-UHFFFAOYSA-N
butane, 1-chloro-4-iodo,tetramethylene chloroiodide,4-chlorobutyl iodide,4-iodobutyl chloride,1-chloro4-iodobutane,4-chloro-1-iodobutane,4-iodo-1-chlorobutane,1-iodo-4-chlorobutane,acmc-1buxg,1-chloro-4-iodo-butane
82527
1-chloro-4-iodobutane
C(CCI)CCl
Infrared Spectrum Authentic
Quantity 100g
Packaging Glass bottle
Refractive Index 1.5380 to 1.5400 (20°C, 589nm)
Color Yellow to Brown
Linear Formula I(CH2)4Cl
Formula Weight 218.46
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Chloro-4-iodobutane

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.