Alfa Aesar™ 1-Chloro-5-trimethylsilyl-4-pentyne, 97%

Manufacturer: Alfa Aesar™ H53393-14

 View more versions of this product

Catalog No. AAH5339314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77113-48-5
    Molecular Formula C8H15ClSi
    Linear Formula Cl(CH2)3C≡CSi(CH3)3
    Formula Weight 174.75g/mol
    MDL Number MFCD00671353
    Synonym (5-Chloro-1-pentynyl)trimethylsilane
    Chemical Name or Material 1-Chloro-5-trimethylsilyl-4-pentyne

    Specifications

    Boiling Point 75° to 77°C (21mmHg)
    Quantity 25g
    Flash Point 65°C (149°F)
    CAS 77113-48-5
    Molecular Formula C8H15ClSi
    Linear Formula Cl(CH2)3C≡CSi(CH3)3
    Formula Weight 174.75g/mol
    MDL Number MFCD00671353
    Synonym (5-Chloro-1-pentynyl)trimethylsilane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1-Chloro-5-trimethylsilyl-4-pentyne
    Assay 97%

    Description

    Applications
    1-Chloro-5-trimethylsilyl-4-pentyne is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit