Alfa Aesar™ 1-Chloroadamantane, 98%

Manufacturer: Alfa Aesar™ 1516922

Quantity 100g

 View more versions of this product

Catalog No. AA1516922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 935-56-8
    Formula Weight 170.68
    MDL Number MFCD00075627
    Synonym 1-Adamantyl chloride
    Chemical Name or Material 1-Chloroadamantane

    Specifications

    Formula Weight 170.68
    Physical Form Crystalline
    MDL Number MFCD00075627
    Melting Point 165°C
    Quantity 100g
    TSCA TSCA
    CAS 935-56-8
    Synonym 1-Adamantyl chloride
    Chemical Name or Material 1-Chloroadamantane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit