Alfa Aesar™ 1-Chloroanthraquinone, 98%

Manufacturer: Alfa Aesar™ B2452222

Quantity 100g

 View more versions of this product

Catalog No. AAB2452222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 82-44-0
    Molecular Formula C14H7ClO2
    Formula Weight 242.66
    MDL Number MFCD00001189
    Beilstein 1912752
    Chemical Name or Material 1-Chloroanthraquinone

    Specifications

    Formula Weight 242.66
    Molecular Formula C14H7ClO2
    MDL Number MFCD00001189
    Health Hazard 1 Warning
    Melting Point 158° to 162°C
    TSCA TSCA
    CAS 82-44-0
    Beilstein 1912752
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CB6150000
    Quantity 100g
    Chemical Name or Material 1-Chloroanthraquinone
    Assay 98%

    Description

    Applications
    It is used as a dyestuff intermediate.

    Solubility
    Soluble in hot toluene. (almost transparency)

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit