Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorobutane, 99.5%, for analysis, Acros Organics

Click to view available options
Quantity:
1L
Packaging:
Glass Bottle
109-69-3
C4H9Cl
92.57
VFWCMGCRMGJXDK-UHFFFAOYSA-N
butyl chloride,n-butyl chloride,butane, 1-chloro,chlorure de butyle,nbc wormer,n-propylcarbinyl chloride,sure shot,butane, chloro,chlorobutane,n-chlorobutane
8005
1-chlorobutane
CCCCCl
109-69-3
C4H9Cl
92.57
VFWCMGCRMGJXDK-UHFFFAOYSA-N
butyl chloride,n-butyl chloride,butane, 1-chloro,chlorure de butyle,nbc wormer,n-propylcarbinyl chloride,sure shot,butane, chloro,chlorobutane,n-chlorobutane
8005
1-chlorobutane
CCCCCl
Viscosity 0.45 mPa.s (20°C)
Specific Gravity 0.88
Packaging Glass Bottle
Flash Point −12°C
Melting Point -123.0°C
Refractive Index 1.4014 to 1.4034
Quantity 1L
Boiling Point 77.0°C to 78.0°C
Density 0.8800g/mL
Assay Percent Range 99.4% min. (GC)
Linear Formula CH3(CH2)3Cl
Merck Index 15,1561
Solubility Information Solubility in water: 0.5g/L (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 92.57
Percent Purity 99.5%
Grade Analysis
Chemical Name or Material 1-Chlorobutane
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 203-696-6