1-Chlorobutane, 99+%

Manufacturer: Alfa Aesar A11494AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11494AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-69-3
    Molecular Formula CH3(CH2)3Cl
    Formula Weight 92.57
    MDL Number MFCD00001009
    UN Number UN1127
    Beilstein 1730909
    Merck Index 14,1560
    Chemical Name or Material 1-Chlorobutane

    Specifications

    Formula Weight 92.57
    Quantity 500mL
    TSCA TSCA
    MDL Number MFCD00001009
    UN Number UN1127
    Beilstein 1730909
    Merck Index 14,1560
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond con
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
    RTECS Number EJ6300000
    Physical Form Liquid
    Density 0.886
    Assay Percent Range >99%
    Molecular Formula CH3(CH2)3Cl
    CAS 109-69-3
    Chemical Name or Material 1-Chlorobutane
    Assay ≥99%

    Description

    Applications
    1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

    Solubility
    Miscible with alcohol and ether. Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond con

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit