Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorobutane, BioSolv(R), 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
109-69-3
109-69-3
Packaging Amber Glass Bottle
Quantity 1 L
Titratable Acid 0.0002 meq/g
Water 100 ppm
CAS Max % 1
Percent Purity 0.995
Grade High Purity Solvent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.