Alfa Aesar™ 1-Chlorohexane, 96%

Manufacturer: Alfa Aesar™ B2237930

Quantity 250g

 View more versions of this product

Catalog No. AAB2237930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 544-10-5
    Molecular Formula C6H13Cl
    Linear Formula CH3(CH2)5Cl
    Formula Weight 120.62
    MDL Number MFCD00001018
    UN Number UN1993
    Beilstein 1731289
    Merck Index 14,2144
    Synonym n-Hexyl chloride
    Chemical Name or Material 1-Chlorohexane

    Specifications

    Formula Weight 120.62
    TSCA TSCA
    Molecular Formula C6H13Cl
    Linear Formula CH3(CH2)5Cl
    MDL Number MFCD00001018
    UN Number UN1993
    Beilstein 1731289
    Merck Index 14,2144
    Synonym n-Hexyl chloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 26°C (78°F)
    Boiling Point 132° to 134°C
    Melting Point −94°C
    Quantity 250g
    CAS 544-10-5
    Chemical Name or Material 1-Chlorohexane
    Assay 96%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit