Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloromethyl-1H-1,2,4-triazole, hydrochloride, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
84387-61-1
C3H4ClN3·ClH
154
GXVWDLWTFIDZLR-UHFFFAOYSA-N
1-chloromethyl-1h-1,2,4-triazole hydrochloride,1-chloromethyl-1,2,4-triazole hydrochloride,1h-1,2,4-triazole,1-chloromethyl-,monohydrochloride,1h-1,2,4-triazole, 1-chloromethyl-, monohydrochloride
13268168
1-(chloromethyl)-1,2,4-triazole;hydrochloride
C1=NN(C=N1)CCl.Cl
84387-61-1
C3H4ClN3·ClH
154
GXVWDLWTFIDZLR-UHFFFAOYSA-N
1-chloromethyl-1h-1,2,4-triazole hydrochloride,1-chloromethyl-1,2,4-triazole hydrochloride,1h-1,2,4-triazole,1-chloromethyl-,monohydrochloride,1h-1,2,4-triazole, 1-chloromethyl-, monohydrochloride
13268168
1-(chloromethyl)-1,2,4-triazole;hydrochloride
C1=NN(C=N1)CCl.Cl
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 154
Percent Purity 98%
Chemical Name or Material 1-Chloromethyl-1H-1,2,4-triazole, hydrochloride