Alfa Aesar™ 1-(Chloromethyl)naphthalene, 94%

Manufacturer: Alfa Aesar™ B2438530

Quantity 250g

 View more versions of this product

Catalog No. AAB2438530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 86-52-2
    Molecular Formula C11H9Cl
    Formula Weight 176.65
    MDL Number MFCD00004042
    UN Number UN3261
    Beilstein 636885
    Synonym 1-Naphthylmethyl chloride
    Chemical Name or Material 1-(Chloromethyl)naphthalene

    Specifications

    Formula Weight 176.65
    Odor Pungent
    Molecular Formula C11H9Cl
    MDL Number MFCD00004042
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Flash Point 132°C (270°F)
    Boiling Point 291° to 292°C
    Quantity 250g
    TSCA TSCA
    CAS 86-52-2
    UN Number UN3261
    Beilstein 636885
    Synonym 1-Naphthylmethyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    RTECS Number QJ2800000
    Melting Point 31° to 33°C
    Chemical Name or Material 1-(Chloromethyl)naphthalene
    Assay 94%

    Description

    Applications
    1-(Chloromethyl)naphthalene is a key material for synthesis dye pigment and as fluorescent brightening agent. It is employed in synthetic resin and medicine. Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives produce ortho- or para-substituted carbocycles in satisfactory to excellent yields.

    Solubility
    Insoluble in water.

    Notes
    Store at room temperature. Incompatible with oxidizing agents and bases.

    GHS H Statement
    H314-H318-H302-H312
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit