Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(Chloromethyl)naphthalene, 94%

Manufacturer:  Alfa Aesar™ B2438536

Quantity 500g

 View more versions of this product

Catalog No. AAB2438536

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
1-(Chloromethyl)naphthalene is a key material for synthesis dye pigment and as fluorescent brightening agent. It is employed in synthetic resin and medicine. Palladium-catalyzed nucleophilic dearomatization of chloromethyl naphthalene derivatives produce ortho- or para-substituted carbocycles in satisfactory to excellent yields.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents and bases.
Description & Specifications

Chemical Identifiers

86-52-2
176.65
UN3261
1-Naphthylmethyl chloride
C11H9Cl
MFCD00004042
636885
1-(Chloromethyl)naphthalene

Specifications

132°C (270°F)
31° to 33°C
86-52-2
C11H9Cl
MFCD00004042
636885
1-(Chloromethyl)naphthalene
291° to 292°C
500g
Pungent
176.65
UN3261
1-Naphthylmethyl chloride
94%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : QJ2800000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit