Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloronaphthalene, 85%, technical, remainder 2-Chloronaphthalene, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
Packaging:
Glass bottle
90-13-1,91-58-7
C10H7Cl
162.62
JTPNRXUCIXHOKM-UHFFFAOYSA-N
alpha-chloronaphthalene,naphthalene, 1-chloro,1-naphthyl chloride,chloronaphthalene,1-chloro-naphthalene,alpha-naphthyl chloride,1-chlornaftalen,1-chloro naphthalene,alpha-chlornaphthalene,.alpha.-chloronaphthalene
7003
1-chloronaphthalene
C1=CC=C2C(=C1)C=CC=C2Cl
90-13-1,91-58-7
C10H7Cl
162.62
JTPNRXUCIXHOKM-UHFFFAOYSA-N
alpha-chloronaphthalene,naphthalene, 1-chloro,1-naphthyl chloride,chloronaphthalene,1-chloro-naphthalene,alpha-naphthyl chloride,1-chlornaftalen,1-chloro naphthalene,alpha-chlornaphthalene,.alpha.-chloronaphthalene
7003
1-chloronaphthalene
C1=CC=C2C(=C1)C=CC=C2Cl
Specific Gravity 1.194
Melting Point -2.5°C
Packaging Glass bottle
Flash Point 121°C
Refractive Index 1.6310 to 1.6340
Density 1.1940g/mL
Infrared Spectrum Authentic
Boiling Point 259.0°C
Quantity 100mL
Color Colorless to Yellow
Assay Percent Range 80% min. (HPLC)
Linear Formula C10H7Cl
Beilstein 05,541
Merck Index 15,2151
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,petroleum,ether and alcohol
Formula Weight 162.62
Physical Form Liquid
Percent Purity 85%
Grade Technical
Chemical Name or Material 1-Chloronaphthalene
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 201-967-3