Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloropentane, 98%

Manufacturer:  Alfa Aesar™ B2473330

Quantity 250g

 View more versions of this product

Catalog No. AAB2473330


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

543-59-9
CH3(CH2)4Cl
MFCD00001015
14,606
1-Chloropentane
C5H11Cl
106.6
UN1107
n-Pentyl chloride; n-Amyl chloride

Specifications

3°C (37°F)
106° to 108°C
250g
C5H11Cl
106.6
UN1107
n-Pentyl chloride; n-Amyl chloride
98%
Sweet
−99°C
543-59-9
CH3(CH2)4Cl
MFCD00001015
14,606
1-Chloropentane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P273-P403
Avoid release to the environment.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit